5-(3-Hydroxyphenyl)-1H-tetrazole 98%

Synonyms: 3-(1H-Tetrazol-5-yl)phenol

BD100838-250MG BD100838-100MG
BLDPBD100838-250MGEA 72.5 EUR
BLDPBD100838-250MG BLDPBD100838-100MG
5-(3-Hydroxyphenyl)-1H-tetrazole 98%
5-(3-Hydroxyphenyl)-1H-tetrazole

Storage condition: Sealed in dry, 2-8°C

Formula: C₇H₆N₄O
MW: 162,16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06797247
CAS Number: 96859-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR