4'-Bromo-2-chloroacetanilide 98%
|
|
Synonyms:
N-(4-Bromophenyl)-2-chloroacetamide
, N1-(4-bromophenyl)-2-chloroacetamide
BLDPBD105824-1GEA
36.8
EUR
BLDPBD105824-1G
BLDPBD105824-250MG
BLDPBD105824-5G
BLDPBD105824-100MG
4'-Bromo-2-chloroacetanilide 98%
4'-Bromo-2-chloroacetanilide
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₈H₇BrClNO MW: 248,5043 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00018903 CAS Number: 2564-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...