2-Methoxy-N-methyaniline 95%

Synonyms: 2-Methoxy-N-methylaniline

BD10679-1G BD10679-10G BD10679-250MG BD10679-100G BD10679-25G BD10679-5G
BLDPBD10679-1GEA 17.9 EUR
BLDPBD10679-1G BLDPBD10679-10G BLDPBD10679-250MG BLDPBD10679-100G BLDPBD10679-25G BLDPBD10679-5G
2-Methoxy-N-methyaniline 95%
2-Methoxy-N-methyaniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₈H₁₁NO
MW: 137,1790 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03093822
CAS Number: 10541-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR