2-Isobutyl-4-methylthiazole 98%

Supplier: BLD PHARMATECH GMBH

BLDPBD10715-25GEA 27.3 EUR
BLDPBD10715-25G BLDPBD10715-5G
2-Isobutyl-4-methylthiazole 98%
2-Isobutyl-4-methylthiazole

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₁₃NS
MW: 155,2605 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00053138
CAS Number: 61323-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR