6,7-Dichloroquinoxaline-2,3-dione 97%

Supplier: BLD PHARMATECH GMBH

Synonyms: 6,7-Dichloroquinoxaline-2,3(1H,4H)-dione6,7-Dichloro-1,4-dihydroquinoxaline-2,3-dione

BLDPBD107822-25GEA 210 EUR
BLDPBD107822-25G BLDPBD107822-5G BLDPBD107822-1G
6,7-Dichloroquinoxaline-2,3-dione 97%
6,7-Dichloroquinoxaline-2,3-dione

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₄Cl₂N₂O₂
MW: 231,0356 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078575
CAS Number: 25983-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR