6-Bromo-4-methoxyquinoline 98%

Supplier: BLD PHARMATECH GMBH

BLDPBD108502-100MGEA 51.5 EUR
BLDPBD108502-100MG BLDPBD108502-1G BLDPBD108502-250MG BLDPBD108502-5G
6-Bromo-4-methoxyquinoline 98%
6-Bromo-4-methoxyquinoline

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₈BrNO
MW: 238,0806 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04036192
CAS Number: 874792-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR