5-Amino-2,4,6-triiodoisophthalic acid 98%

Supplier: BLD PHARMATECH GMBH

BLDPBD10851-10GEA 8.4 EUR
BLDPBD10851-10G BLDPBD10851-100G BLDPBD10851-500G BLDPBD10851-1000G BLDPBD10851-1G BLDPBD10851-5G BLDPBD10851-25G
5-Amino-2,4,6-triiodoisophthalic acid 98%
5-Amino-2,4,6-triiodoisophthalic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₈H₄I₃NO₄
MW: 558,84 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00190167
CAS Number: 35453-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR