6-Chloro-3-iodoimidazo[1,2-a]pyridazine 96%

Synonyms: 6-Chloro-3-iodoimidazo[1,2-b]pyridazine

BD108808-250MG BD108808-10G BD108808-5G BD108808-1G BD108808-100MG
BLDPBD108808-250MGEA 47.3 EUR
BLDPBD108808-250MG BLDPBD108808-10G BLDPBD108808-5G BLDPBD108808-1G BLDPBD108808-100MG
6-Chloro-3-iodoimidazo[1,2-a]pyridazine 96%
6-Chloro-3-iodoimidazo[1,2-a]pyridazine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₆H₃ClIN₃
MW: 279,4700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11044749
CAS Number: 923595-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR