6-Chloro-4-methoxy indole 97%

Supplier: BLD PHARMATECH GMBH

Synonyms: 6-Chloro-4-methoxy-1H-indole

BLDPBD108841-100MGEA 137 EUR
BLDPBD108841-100MG BLDPBD108841-250MG
6-Chloro-4-methoxy indole 97%
6-Chloro-4-methoxy indole

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₈ClNO
MW: 181,619 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00238560
CAS Number: 117970-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR