Dioctyl malonate 95%

Synonyms: Dioctylmalonate

BD1112-1G BD1112-5G BD1112-25G BD1112-250MG
BLDPBD1112-1GEA 38.9 EUR
BLDPBD1112-1G BLDPBD1112-5G BLDPBD1112-25G BLDPBD1112-250MG
Dioctyl malonate 95%
Dioctyl malonate
Formula: C₁₉H₃₆O₄
MW: 328,487 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00048936
CAS Number: 16958-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR