8-Methylimidazo[1,2-a]pyridine 95%
|
|
BLDPBD111425-250MGEA
14.7
EUR
BLDPBD111425-250MG
BLDPBD111425-100MG
BLDPBD111425-1G
BLDPBD111425-5G
8-Methylimidazo[1,2-a]pyridine 95%
8-Methylimidazo[1,2-a]pyridine
Storage condition: Sealed in dry, 2-8°C
Formula:
C₈H₈N₂ MW: 132,1600 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD05663805 CAS Number: 874-10-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...