Potassium acetate 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: Acetic acid potassium salt

BLDPBD112157-100GEA 8.4 EUR
BLDPBD112157-100G BLDPBD112157-1000G BLDPBD112157-25G BLDPBD112157-500G
Potassium acetate 98%
Potassium acetate

Storage condition: Inert atmosphere, Room Temperature

Formula: CH₃COOK
MW: 98,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR