Amino-naphthalen-2-yl-acetic acid 95%

Supplier: BLD PHARMATECH GMBH

Synonyms: 2-Amino-2-(naphthalen-2-yl)acetic acid

BLDPBD113997-100MGEA 126 EUR
BLDPBD113997-100MG BLDPBD113997-250MG
Amino-naphthalen-2-yl-acetic acid 95%
Amino-naphthalen-2-yl-acetic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₁₁NO₂
MW: 201,221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02662462
CAS Number: 33741-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR