6-O-Palmitoyl-L-ascorbic acid 95%

Synonyms: L-Ascorbic acid 6-palmitate,Palmitoyl-L-ascorbic acid,Ascorbyl Palmitate,(S)-2-((R)-3,4-Dihydroxy-5-oxo-2,5-dihydrofuran-2-yl)-2-hydroxyethyl palmitate,L(+)-Ascorbyl palmitate

BD114742-10G BD114742-100G BD114742-25G BD114742-500G BD114742-1000G
BLDPBD114742-10GEA 7.4 EUR
BLDPBD114742-10G BLDPBD114742-100G BLDPBD114742-25G BLDPBD114742-500G BLDPBD114742-1000G
6-O-Palmitoyl-L-ascorbic acid 95%
6-O-Palmitoyl-L-ascorbic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₂₂H₃₈O₇
MW: 414,54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005377
CAS Number: 137-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR