2,4,6-Trimethylbenzenesulphonamide 96%
|
|
BLDPBD115570-5GEA
49.4
EUR
BLDPBD115570-5G
BLDPBD115570-25G
BLDPBD115570-0.025
BLDPBD115570-1G
2,4,6-Trimethylbenzenesulphonamide 96%
2,4,6-Trimethylbenzenesulphonamide
Storage condition: Sealed in dry, Room Temperature
Formula:
C₉H₁₃NO₂S MW: 199,2700 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00463245 CAS Number: 4543-58-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...