Difurfuryl sulphide 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: Bis(furan-2-ylmethyl)sulfane

BLDPBD11833-100GEA 62 EUR
BLDPBD11833-100G BLDPBD11833-5G BLDPBD11833-1G BLDPBD11833-25G
Difurfuryl sulphide 98%
Difurfuryl sulphide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₀H₁₀O₂S
MW: 194,2500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010081
CAS Number: 13678-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR