Calcium hydrogen orthophosphate 99%

Synonyms: Calcium phosphate dibasicAnhydrous Dibasic Calcium PhosphateCalcium hydrogen phosphateCalcium perphosphate, ammonifiedCalcium phosphate, dibasic

BLDPBD119735-25GEA 6.3 EUR
BLDPBD119735-25G BLDPBD119735-100G BLDPBD119735-0.025 BLDPBD119735-1000G BLDPBD119735-500G
Calcium hydrogen orthophosphate 99%
Calcium hydrogen orthophosphate

Storage condition: Inert atmosphere, Room Temperature

Formula: CaHPO₄
MW: 136,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010909
CAS Number: 7757-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR