4-Propoxy-3-methylphenylboronic acid 97%

Synonyms: (3-Methyl-4-propoxyphenyl)boronic acid3-Methyl-4-propoxybenzeneboronic acid

BLDPBD12256-250MGEA 0 EUR
BLDPBD12256-250MG BLDPBD12256-1G BLDPBD12256-5G
4-Propoxy-3-methylphenylboronic acid 97%
4-Propoxy-3-methylphenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₀H₁₅BO₃
MW: 194,0400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06659872
CAS Number: 279262-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR