6-Fluorochromone-2-carboxylic acid 98%
Supplier: BLD PHARMATECH GMBH
|
|
Synonyms:
6-Fluoro-4-oxo-4H-chromene-2-carboxylic acid
BLDPBD12489-1GEA
33.6
EUR
BLDPBD12489-1G
BLDPBD12489-250MG
BLDPBD12489-5G
6-Fluorochromone-2-carboxylic acid 98%
6-Fluorochromone-2-carboxylic acid
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₀H₅FO₄ MW: 208,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03070543 CAS Number: 99199-59-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...