Disodium hydrogen phosphate dodecahydrate 99%

Supplier: BLD PHARMATECH GMBH

Synonyms: Dibasic sodium phosphate dodecahydrateSodium phosphate dibasic dodecahydrateSodium hydrogen phosphate dodecahydratedi-Sodium hydrogen orthophosphate dodecahydrateSodium monohydrogen phosphate dodecahydrate

BLDPBD126089-1000GEA 16.8 EUR
BLDPBD126089-1000G BLDPBD126089-500G BLDPBD126089-100G
Disodium hydrogen phosphate dodecahydrate 99%
di-Sodium hydrogen phosphate dodecahydrate

Storage condition: Sealed in dry, Room Temperature

Formula: Na₂HPO₄·12H₂O
MW: 358,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149181
CAS Number: 10039-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR