Ethyl glycolate 97%

Synonyms: Ethyl Hydroxyacetate,Hydroxyacetic Acid Ethyl Ester,Glycolic Acid Ethyl Ester,Ethyl 2-hydroxyacetate

BD129346-500G BD129346-25G BD129346-100G BD129346-5G BD129346-1000G BD129346-10G BD129346-1G
BLDPBD129346-500GEA 114 EUR
BLDPBD129346-500G BLDPBD129346-25G BLDPBD129346-100G BLDPBD129346-5G BLDPBD129346-1000G BLDPBD129346-10G BLDPBD129346-1G
Ethyl glycolate 97%
Ethyl glycolate

Storage condition: Sealed in dry, Room Temperature

Formula: C₄H₈O₃
MW: 104,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00021970
CAS Number: 623-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR