1-Hydroxy-4-(p-toluidino)-9,10-anthraquinone 97%

Synonyms: 1-Hydroxy-4-(p-tolylamino)anthracene-9,10-dione

BD1301-500G BD1301-100G BD1301-25G BD1301-1000G
BLDPBD1301-500GEA 86.1 EUR
BLDPBD1301-500G BLDPBD1301-100G BLDPBD1301-25G BLDPBD1301-1000G
1-Hydroxy-4-(p-toluidino)-9,10-anthraquinone 97%
1-Hydroxy-4-(p-toluidino)-9,10-anthraquinone

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₂₁H₁₅NO₃
MW: 329,3500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00019148
CAS Number: 81-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR