Isopropyltrichlorosilane 95%

Synonyms: Trichloro(isopropyl)silane

BLDPBD135340-1GEA 48.3 EUR
BLDPBD135340-1G BLDPBD135340-250MG BLDPBD135340-5G
Isopropyltrichlorosilane 95%
Isopropyltrichlorosilane
Formula: C₃H₇Cl₃Si
MW: 177,5322 g/mol
CAS Number: 4170-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR