1-Bromo-4-hexyloxybenzene 97%

Supplier: BLD PHARMATECH GMBH

Synonyms: 1-Bromo-4-(hexyloxy)benzene1-(4-Bromophenoxy)hexane

BLDPBD14176-5GEA 43.1 EUR
BLDPBD14176-5G BLDPBD14176-1G BLDPBD14176-25G
1-Bromo-4-hexyloxybenzene 97%
1-Bromo-4-hexyloxybenzene

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₁₇BrO
MW: 257,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173753
CAS Number: 30752-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR