2-Acetamidoethanol 90%

Synonyms: N-Acetylethanolamine

BD143005-5G BD143005-25G
BLDPBD143005-5GEA 8.4 EUR
BLDPBD143005-5G BLDPBD143005-25G
2-Acetamidoethanol 90%
2-Acetamidoethanol

Storage condition: Sealed in dry, Room Temperature

Formula: C₄H₉NO₂
MW: 103,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002836
CAS Number: 142-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR