Sodium hexanoate 99%

Supplier: BLD PHARMATECH GMBH

Synonyms: N-caproicacidsodiumsalt

BLDPBD143821-100GEA 175 EUR
BLDPBD143821-100G BLDPBD143821-25G BLDPBD143821-5G
Sodium hexanoate 99%
Sodium hexanoate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₆H₁₁NaO₂
MW: 138,1401 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059056
CAS Number: 10051-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR