Perinaphthenone 98%

Synonyms: 1H-Phenalen-1-one

BLDPBD147319-1GEA 153 EUR
BLDPBD147319-1G BLDPBD147319-250MG
Perinaphthenone 98%
Perinaphthenone

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₃H₈O
MW: 180,202 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004143
CAS Number: 548-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR