Potassium-3-fluoro-4-methoxyphenyltrifluoroborate 95%
|
|
Synonyms:
Potassium trifluoro(3-fluoro-4-methoxyphenyl)borate
, Potassium(3-Fluoro-4-methoxyphenyl)trifluoroborate
BLDPBD148664-1GEA
23.1
EUR
BLDPBD148664-1G
BLDPBD148664-250MG
BLDPBD148664-5G
Potassium-3-fluoro-4-methoxyphenyltrifluoroborate 95%
Potassium 3-fluoro-4-methoxyphenyltrifluoroborate
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₇H₆BF₄KO MW: 232,0300 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04115757 CAS Number: 850623-62-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...