Potassium-3,5-bis(trifluoromethyl)phenyltrifluoroborate 97%

Synonyms: Potassium (3,5-bis(trifluoromethyl)phenyl)trifluoroborate

BD148706-1G BD148706-250MG BD148706-5G
BLDPBD148706-1GEA 14.7 EUR
BLDPBD148706-1G BLDPBD148706-250MG BLDPBD148706-5G
Potassium-3,5-bis(trifluoromethyl)phenyltrifluoroborate 97%
Potassium 3,5-bis(trifluoromethyl)phenyltrifluoroborate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₃BF₉K
MW: 320,01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03701625
CAS Number: 166328-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR