Potassium-3-formylphenyltrifluoroborate 97%

Synonyms: Potassium trifluoro(3-formylphenyl)borate

BLDPBD148709-5GEA 117 EUR
BLDPBD148709-5G BLDPBD148709-1G
Potassium-3-formylphenyltrifluoroborate 97%
Potassium 3-formylphenyltrifluoroborate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₅BF₃KO
MW: 212,02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07784378
CAS Number: 871231-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR