Potassium-3-hydroxyphenyltrifluoroborate 97%
|
|
Synonyms:
Potassium trifluoro(3-hydroxyphenyl)borate
BLDPBD148710-1GEA
15.8
EUR
BLDPBD148710-1G
BLDPBD148710-5G
Potassium-3-hydroxyphenyltrifluoroborate 97%
Potassium 3-hydroxyphenyltrifluoroborate
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₆H₅BF₃KO MW: 200,0100 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07784422 CAS Number: 871231-45-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...