Potassium iodide 99%
BLDPBD148824-100GEA
15.8
EUR
BLDPBD148824-100G
BLDPBD148824-10G
BLDPBD148824-25G
BLDPBD148824-500G
BLDPBD148824-1000G
Potassium iodide 99%
Potassium iodide
Formula:
Kl MW: 166 g/mol |
MDL Number:
MFCD00011405 CAS Number: 7681-11-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...