4-Fluoro-2-methylphenylboronic acid pinacol ester 98%
|
|
Synonyms:
2-(4-Fluoro-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
, 4-Fluoro-2-methylbenzeneboronic acid pinacol ester
BLDPBD14885-1GEA
9.5
EUR
BLDPBD14885-1G
4-Fluoro-2-methylphenylboronic acid pinacol ester 98%
4-Fluoro-2-methylphenylboronic acid pinacol ester
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₃H₁₈BFO₂ MW: 236,09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05863911 CAS Number: 815631-56-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...