4-(Methylthio)phenylboronic acid pinacol ester 98%

Synonyms: 4,4,5,5-Tetramethyl-2-(4-(methylthio)phenyl)-1,3,2-dioxaborolane

BD14942-5G BD14942-1G BD14942-250MG
BLDPBD14942-5GEA 133 EUR
BLDPBD14942-5G BLDPBD14942-1G BLDPBD14942-250MG
4-(Methylthio)phenylboronic acid pinacol ester 98%
4-(Methylthio)phenylboronic acid pinacol ester

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₃H₁₉BO₂S
MW: 250,1648 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05155222
CAS Number: 190788-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR