4-(4-Bromobenzyloxy)benzaldehyde 98%

Synonyms: 4-((4-Bromobenzyl)oxy)benzaldehyde

BD15193-1G BD15193-250MG BD15193-5G BD15193-100MG
BLDPBD15193-1GEA 224 EUR
BLDPBD15193-1G BLDPBD15193-250MG BLDPBD15193-5G BLDPBD15193-100MG
4-(4-Bromobenzyloxy)benzaldehyde 98%
4-(4-Bromobenzyloxy)benzaldehyde

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₄H₁₁BrO₂
MW: 291,1399 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02091001
CAS Number: 149833-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR