tert-Butyl 4-hydroxybenzoate 96%

Supplier: BLD PHARMATECH GMBH

BLDPBD152937-100GEA 355 EUR
BLDPBD152937-100G BLDPBD152937-1G BLDPBD152937-10G BLDPBD152937-0.025 BLDPBD152937-5G BLDPBD152937-250MG BLDPBD152937-25G
tert-Butyl 4-hydroxybenzoate 96%
tert-Butyl 4-hydroxybenzoate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₁H₁₄O₃
MW: 194,227 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09953072
CAS Number: 25804-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR