2-(Methoxycarbonyl)phenylboronic acid pinacol ester 97%

Synonyms: 2-(2-Methoxycarbonylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid methyll ester,2-(Methoxycarbonyl)phenylboronic acid Pinacol ester,Methyl 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

BD15548-250MG BD15548-1G BD15548-5G
BLDPBD15548-250MGEA 6.3 EUR
BLDPBD15548-250MG BLDPBD15548-1G BLDPBD15548-5G
2-(Methoxycarbonyl)phenylboronic acid pinacol ester 97%
2-(Methoxycarbonyl)phenylboronic acid pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₄H₁₉BO₄
MW: 262,1093 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05663866
CAS Number: 653589-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR