3-Amino-1-ethyl-1H-pyrazole 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: 1-Ethyl-1H-pyrazol-3-amine

BLDPBD157631-100MGEA 55.7 EUR
BLDPBD157631-100MG BLDPBD157631-250MG BLDPBD157631-1G
3-Amino-1-ethyl-1H-pyrazole 98%
3-Amino-1-ethyl-1H-pyrazole

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₅H₉N₃
MW: 111,1451 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03422571
CAS Number: 55361-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR