3-Bromo-2-methylthiophene 95%

BLDPBD158060-25GEA 408 EUR
BLDPBD158060-25G BLDPBD158060-1G
3-Bromo-2-methylthiophene 95%
3-Bromo-2-methylthiophene

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formula: C₅H₅BrS
MW: 177,0622 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09033008
CAS Number: 30319-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR