3-Fluoro-2-nitroaniline 98%

BLDPBD158085-10GEA 56.7 EUR
BLDPBD158085-10G BLDPBD158085-100G BLDPBD158085-5G BLDPBD158085-25G BLDPBD158085-500G BLDPBD158085-1G
3-Fluoro-2-nitroaniline 98%
3-Fluoro-2-nitroaniline

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₆H₅FN₂O₂
MW: 156,1145 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07368750
CAS Number: 567-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR