2-[(2-Bromoacetyl)amino]benzophenone 95%

Synonyms: N-(2-Benzoylphenyl)-2-bromoacetamide

BD15833-100MG BD15833-1G BD15833-250MG
BLDPBD15833-100MGEA 73.5 EUR
BLDPBD15833-100MG BLDPBD15833-1G BLDPBD15833-250MG
2-[(2-Bromoacetyl)amino]benzophenone 95%
2-[(2-Bromoacetyl)amino]benzophenone

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₅H₁₂BrNO₂
MW: 318,1653 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00277799
CAS Number: 14439-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR