4-Bromo-2-(trifluoromethoxy)benzaldehyde 97%

Supplier: BLD PHARMATECH GMBH

BLDPBD158496-1GEA 16.8 EUR
BLDPBD158496-1G BLDPBD158496-250MG BLDPBD158496-0.025 BLDPBD158496-25G BLDPBD158496-5G BLDPBD158496-100G
4-Bromo-2-(trifluoromethoxy)benzaldehyde 97%
4-Bromo-2-(trifluoromethoxy)benzaldehyde

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₄BrF₃O₂
MW: 269,0200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12406886
CAS Number: 220996-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR