2-Bromo-3-chloroimidazo[1,2-a]pyridine 95%

BLDPBD158533-250MGEA 128 EUR
BLDPBD158533-250MG BLDPBD158533-100MG
2-Bromo-3-chloroimidazo[1,2-a]pyridine 95%
2-Bromo-3-chloroimidazo[1,2-a]pyridine

Storage condition: Inert atmosphere, 2-8°C

Formula: C₇H₄BrClN₂
MW: 231,4800 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11869753
CAS Number: 1159511-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR