4-Chloro-3-(tert-butylcarbamoyl)phenylboronic acid 97%

Synonyms: (3-(tert-Butylcarbamoyl)-4-chlorophenyl)boronic acid

BD158594-5G BD158594-1G BD158594-250MG
BLDPBD158594-5GEA 431 EUR
BLDPBD158594-5G BLDPBD158594-1G BLDPBD158594-250MG
4-Chloro-3-(tert-butylcarbamoyl)phenylboronic acid 97%
4-Chloro-3-(tert-butylcarbamoyl)phenylboronic acid
Formula: C₁₁H₁₅BClNO₃
MW: 255,5100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07783864
CAS Number: 871332-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR