2,4-Dimethoxy-4'-hydroxybenzophenone 97%
|
|
Synonyms:
(2,4-Dimethoxyphenyl)(4-hydroxyphenyl)methanone
BLDPBD15943-1GEA
7.4
EUR
BLDPBD15943-1G
BLDPBD15943-5G
BLDPBD15943-100G
BLDPBD15943-10G
BLDPBD15943-25G
2,4-Dimethoxy-4'-hydroxybenzophenone 97%
2,4-Dimethoxy-4'-hydroxybenzophenone
Storage condition: Sealed in dry, 2-8°C
Formula:
C₁₅H₁₄O₄ MW: 258,27 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00137850 CAS Number: 41351-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...