2-Amino-6-bromobenzonitrile 97%

BLDPBD159757-250MGEA 7.4 EUR
BLDPBD159757-250MG BLDPBD159757-10G BLDPBD159757-100G BLDPBD159757-25G BLDPBD159757-1G BLDPBD159757-5G
2-Amino-6-bromobenzonitrile 97%
2-Amino-6-bromobenzonitrile

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₇H₅BrN₂
MW: 197,02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09834769
CAS Number: 77326-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR