2-Bromoimidazo[1,2-a]pyridine 97%
|
|
BLDPBD159795-25GEA
752
EUR
BLDPBD159795-25G
BLDPBD159795-1G
BLDPBD159795-100MG
BLDPBD159795-5G
BLDPBD159795-250MG
2-Bromoimidazo[1,2-a]pyridine 97%
2-Bromoimidazo[1,2-a]pyridine
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₇H₅BrN₂ MW: 197,0300 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09834778 CAS Number: 112581-95-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...