2,3-Difluoroisonicotinic acid 96%
|
|
Synonyms:
2,3-Difluoropyridine-4-carboxylic acid
BLDPBD159944-1GEA
24.2
EUR
BLDPBD159944-1G
BLDPBD159944-5G
BLDPBD159944-25G
BLDPBD159944-250MG
2,3-Difluoroisonicotinic acid 96%
2,3-Difluoroisonicotinic acid
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₆H₃F₂NO₂ MW: 159,09 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09031174 CAS Number: 851386-31-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...