5-Bromo-1,3-dichloro-2-fluorobenzene 98%

Synonyms: 3,5-Dichloro-4-fluorobromobenzene

BD160145-10G BD160145-25G BD160145-5G BD160145-500G BD160145-1G BD160145-100G
BLDPBD160145-10GEA 15.8 EUR
BLDPBD160145-10G BLDPBD160145-25G BLDPBD160145-5G BLDPBD160145-500G BLDPBD160145-1G BLDPBD160145-100G
5-Bromo-1,3-dichloro-2-fluorobenzene 98%
5-Bromo-1,3-dichloro-2-fluorobenzene

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₂BrCl₂F
MW: 243,8900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09996901
CAS Number: 17318-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR