4-Bromo-2-iodobenzoic acid 97%

Supplier: BLD PHARMATECH GMBH

BLDPBD160541-10GEA 60.9 EUR
BLDPBD160541-10G BLDPBD160541-500G BLDPBD160541-100G BLDPBD160541-5G BLDPBD160541-25G BLDPBD160541-1G BLDPBD160541-0.5G
4-Bromo-2-iodobenzoic acid 97%
4-Bromo-2-iodobenzoic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₇H₄BrIO₂
MW: 326,914 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12767500
CAS Number: 1133123-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR